Product Details

SNP ID
rs146788972
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:28572698 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACATTCTTGACAGAACTTTTCCC[C/T]GTGATAACCACCGTATCTCAGCATG
Phenotype
MIM: 615254
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZBED9 PubMed Links

Gene Details

Gene
ZBED9
Gene Name
zinc finger BED-type containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052923.1 3205 Missense Mutation CAG,CGG Q1064R NP_443155.1
XM_011514283.2 3205 Missense Mutation CAG,CGG Q913R XP_011512585.1
XM_011514284.2 3205 Missense Mutation CAG,CGG Q913R XP_011512586.1
XM_011514285.2 3205 Missense Mutation CAG,CGG Q913R XP_011512587.1
XM_011514286.2 3205 Missense Mutation CAG,CGG Q913R XP_011512588.1
XM_011514287.2 3205 Missense Mutation CAG,CGG Q913R XP_011512589.1
XM_011514288.2 3205 Missense Mutation CAG,CGG Q868R XP_011512590.1
XM_017010231.1 3205 Missense Mutation CAG,CGG Q913R XP_016865720.1

View Full Product Details