Product Details

SNP ID
rs147895073
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:167825503 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACTCTGCAGTCCGTTCTCTACGT[C/G]TGCGAAGAATTGGCAGATAACGATG
Phenotype
MIM: 159559
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AFDN PubMed Links

Gene Details

Gene
AFDN
Gene Name
afadin, adherens junction formation factor
There are no transcripts associated with this gene.

Gene
AFDN-AS1
Gene Name
AFDN antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details