Product Details

SNP ID
rs148272640
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:35577043 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGAACATGTTGGCGTATATCCTG[C/T]GGTCCCGCTCGTTGTGCTCCTTGGC
Phenotype
MIM: 602623
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FKBP5 PubMed Links

Gene Details

Gene
FKBP5
Gene Name
FK506 binding protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145775.2 1375 Missense Mutation CAC,CGC H406R NP_001139247.1
NM_001145776.1 1375 Missense Mutation CAC,CGC H406R NP_001139248.1
NM_001145777.1 1375 Intron NP_001139249.1
NM_004117.3 1375 Missense Mutation CAC,CGC H406R NP_004108.1

View Full Product Details