Product Details

SNP ID
rs148586207
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32129291 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCACCTTGACACATGGACTCCA[A/G]GCATTTCTCTATGAGCTCCCCCCAA
Phenotype
MIM: 600984
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATF6B PubMed Links

Gene Details

Gene
ATF6B
Gene Name
activating transcription factor 6 beta
There are no transcripts associated with this gene.

Gene
FKBPL
Gene Name
FK506 binding protein like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022110.3 760 Silent Mutation CTG,TTG L164L NP_071393.2

View Full Product Details