Product Details

SNP ID
rs148871760
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:87410389 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTAGCACCATATTCAAGTTTGATG[C/T]TTGTTGCTTCCTTTTTTCAACCTCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CFAP206 PubMed Links

Gene Details

Gene
CFAP206
Gene Name
cilia and flagella associated protein 206
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031743.2 Intron NP_001026913.1

View Full Product Details