Product Details

SNP ID
rs149099679
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:87477528 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATAAAGTTATTGCTAAGTGTGGG[A/G]ATTTTAGCTAAGTGAGTATAAATAC
Phenotype
MIM: 605634
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC35A1 PubMed Links

Gene Details

Gene
SLC35A1
Gene Name
solute carrier family 35 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168398.1 262 Silent Mutation GGA,GGG G61G NP_001161870.1
NM_006416.4 262 Silent Mutation GGA,GGG G61G NP_006407.1

View Full Product Details