Product Details

SNP ID
rs149186054
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:105278348 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTCAGGGAGTGAAGCGGGACCAC[A/G]CGGTCATCATGGTCAGCAGTGAGGA
Phenotype
MIM: 600400
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PREP PubMed Links

Gene Details

Gene
PREP
Gene Name
prolyl endopeptidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002726.4 2162 Silent Mutation CGC,CGT R643R NP_002717.3
XM_005267044.2 2162 Intron XP_005267101.1
XM_011535925.2 2162 Intron XP_011534227.1

View Full Product Details