Product Details

SNP ID
rs149350645
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30571500 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTGCCACCGCGATGCCGAAGGCG[C/T]CCAAGCAGCAGCCGCCGGAGCCCGA
Phenotype
MIM: 603429
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABCF1 PubMed Links

Gene Details

Gene
ABCF1
Gene Name
ATP binding cassette subfamily F member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025091.1 108 Missense Mutation CCC,TCC P5S NP_001020262.1
NM_001090.2 108 Missense Mutation CCC,TCC P5S NP_001081.1
Gene
PRR3
Gene Name
proline rich 3
There are no transcripts associated with this gene.

View Full Product Details