Product Details

SNP ID
rs149495517
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:118464625 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATAAAACATATAAATAAAAAATT[A/G]TAAATAGTAAGGAACATGTTAAAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEP85L PubMed Links

Gene Details

Gene
CEP85L
Gene Name
centrosomal protein 85 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042475.2 4377 UTR 3 NP_001035940.1
NM_001178035.1 4377 UTR 3 NP_001171506.1
NM_206921.2 4377 Intron NP_996804.2
XM_005266970.1 4377 UTR 3 XP_005267027.1
XM_005266971.1 4377 UTR 3 XP_005267028.1
XM_005266972.4 4377 UTR 3 XP_005267029.1
XM_006715475.3 4377 UTR 3 XP_006715538.1
XM_011535809.2 4377 UTR 3 XP_011534111.1
XM_011535810.2 4377 UTR 3 XP_011534112.1
XM_011535811.1 4377 UTR 3 XP_011534113.1
XM_017010846.1 4377 UTR 3 XP_016866335.1
XM_017010847.1 4377 UTR 3 XP_016866336.1

View Full Product Details