Product Details

SNP ID
rs149642297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:125751808 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCTTCTTATTTCATAGGCAAAGT[A/G]CTAGCTCTGTGATTAGATTGAATTC
Phenotype
MIM: 604674
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HEY2 PubMed Links

Gene Details

Gene
HEY2
Gene Name
hes related family bHLH transcription factor with YRPW motif 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012259.2 143 Missense Mutation ACT,GCT T31A NP_036391.1
XM_017010627.1 143 UTR 5 XP_016866116.1
XM_017010628.1 143 UTR 5 XP_016866117.1
XM_017010629.1 143 UTR 5 XP_016866118.1
Gene
LOC105377986
Gene Name
uncharacterized LOC105377986
There are no transcripts associated with this gene.

View Full Product Details