Product Details

SNP ID
rs150175763
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:96572422 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAGAGCAATTCCACCTCCACTTG[A/G]CCCCACAGGAAAAAGTGTTAACAGC
Phenotype
MIM: 605126
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FHL5 PubMed Links

Gene Details

Gene
FHL5
Gene Name
four and a half LIM domains 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170807.2 Intron NP_001164278.1
NM_001322466.1 Intron NP_001309395.1
NM_001322467.1 Intron NP_001309396.1
NM_020482.5 Intron NP_065228.4

View Full Product Details