Product Details

SNP ID
rs150278073
Assay Type
Functionally Tested
NCBI dbSNP Submissions
2
Location
Chr.6:31111455 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTTCCAAGAAGAGCCAGGAGGGC[A/G]GAGGACTCCAGGAGGAAATGGGTTA
Phenotype
MIM: 611401 MIM: 602593 MIM: 613525
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
C6orf15 PubMed Links

Gene Details

Gene
C6orf15
Gene Name
chromosome 6 open reading frame 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014070.2 904 Missense Mutation CGC,TGC R302C NP_054789.2
Gene
CDSN
Gene Name
corneodesmosin
There are no transcripts associated with this gene.

Gene
PSORS1C1
Gene Name
psoriasis susceptibility 1 candidate 1
There are no transcripts associated with this gene.

View Full Product Details