Product Details

SNP ID
rs17189092
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30562096 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCTCTTATCAAGAATACCTGCCC[A/G]CCCAAGGATGACCCCCAGGTTATGG
Phenotype
MIM: 603429 MIM: 143024
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABCF1 PubMed Links

Gene Details

Gene
ABCF1
Gene Name
ATP binding cassette subfamily F member 1
There are no transcripts associated with this gene.

Gene
GNL1
Gene Name
G protein nucleolar 1 (putative)
There are no transcripts associated with this gene.

Gene
PRR3
Gene Name
proline rich 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077497.2 1005 Silent Mutation CCA,CCG P123P NP_001070965.1
NM_025263.3 1005 Silent Mutation CCA,CCG P144P NP_079539.2

View Full Product Details