Product Details

SNP ID
rs150474276
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:159767064 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTGCAGGAATTCCCTACTCTGTT[C/T]CAGCATGGAGCTGCCAGATGATCTG
Phenotype
MIM: 100678 MIM: 147460
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACAT2 PubMed Links

Gene Details

Gene
ACAT2
Gene Name
acetyl-CoA acetyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303253.1 381 Missense Mutation CCA,TCA P113S NP_001290182.1
NM_005891.2 381 Missense Mutation CCA,TCA P84S NP_005882.2
Gene
LOC100129518
Gene Name
uncharacterized LOC100129518
There are no transcripts associated with this gene.

Gene
SOD2
Gene Name
superoxide dismutase 2, mitochondrial
There are no transcripts associated with this gene.

View Full Product Details