Product Details
- SNP ID
-
rs150632959
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:50043760 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATCGACCTCCAATCGCTGTACATG[A/G]CTTCCACCTACTAAAGGTGATATGG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB112
PubMed Links
Gene Details
- Gene
- DEFB112
- Gene Name
- defensin beta 112
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001037498.1 |
157 |
Missense Mutation |
CCA,TCA |
P53S |
NP_001032587.1 |
View Full Product Details