Product Details

SNP ID
rs150953346
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:118815347 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGTAGATGTTAACTTTCCTGGA[C/T]GCTTCACCGGCAAGGCTGCGTCTCT
Phenotype
MIM: 610098
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MCM9 PubMed Links

Gene Details

Gene
MCM9
Gene Name
minichromosome maintenance 9 homologous recombination repair factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017696.2 2924 Missense Mutation CAT,CGT H970R NP_060166.2
NM_153255.4 2924 Intron NP_694987.1

View Full Product Details