Product Details

SNP ID
rs150967731
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:28995953 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCTTCTCCCCGGTGTGGATAAGC[C/T]GATGCATACAGAGACGGTTCCTGGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HCG16 PubMed Links

Gene Details

Gene
HCG16
Gene Name
HLA complex group 16 (non-protein coding)
There are no transcripts associated with this gene.

Gene
ZNF311
Gene Name
zinc finger protein 311
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010877.3 1512 Missense Mutation CAG,CGG Q350R NP_001010877.2
NM_001318534.1 1512 Missense Mutation CAG,CGG Q258R NP_001305463.1
NM_001318535.1 1512 Missense Mutation CAG,CGG Q258R NP_001305464.1
XM_005249019.3 1512 Missense Mutation CAG,CGG Q383R XP_005249076.1
XM_006715067.2 1512 Missense Mutation CAG,CGG Q383R XP_006715130.1
XM_011514510.1 1512 Missense Mutation CAG,CGG Q383R XP_011512812.1
XM_011514511.2 1512 Missense Mutation CAG,CGG Q383R XP_011512813.1
XM_011514512.1 1512 Missense Mutation CAG,CGG Q375R XP_011512814.1
XM_011514513.1 1512 Missense Mutation CAG,CGG Q374R XP_011512815.1
XM_011514514.1 1512 Missense Mutation CAG,CGG Q358R XP_011512816.1
XM_011514516.1 1512 Missense Mutation CAG,CGG Q302R XP_011512818.1
XM_011514517.1 1512 Missense Mutation CAG,CGG Q258R XP_011512819.1
XM_011514518.1 1512 Missense Mutation CAG,CGG Q258R XP_011512820.1
XM_017010756.1 1512 Missense Mutation CAG,CGG Q383R XP_016866245.1
XM_017010757.1 1512 Missense Mutation CAG,CGG Q383R XP_016866246.1
XM_017010758.1 1512 Missense Mutation CAG,CGG Q350R XP_016866247.1
XM_017010759.1 1512 Missense Mutation CAG,CGG Q350R XP_016866248.1
XM_017010760.1 1512 Missense Mutation CAG,CGG Q302R XP_016866249.1
XM_017010761.1 1512 Missense Mutation CAG,CGG Q258R XP_016866250.1

View Full Product Details