Product Details

SNP ID
rs151049961
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33263546 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCTTTGCTGTCATCTGCAGCCC[A/G]CTCCTAAGGGAAGACAAAGGGAAAT
Phenotype
MIM: 180473 MIM: 603443
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HCG25 PubMed Links

Gene Details

Gene
HCG25
Gene Name
HLA complex group 25 (non-protein coding)
There are no transcripts associated with this gene.

Gene
RPS18
Gene Name
ribosomal protein S18
There are no transcripts associated with this gene.

Gene
VPS52
Gene Name
VPS52, GARP complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289174.1 1957 Missense Mutation CGG,TGG R511W NP_001276103.1
NM_001289175.1 1957 Missense Mutation CGG,TGG R453W NP_001276104.1
NM_001289176.1 1957 Missense Mutation CGG,TGG R389W NP_001276105.1
NM_022553.5 1957 Missense Mutation CGG,TGG R578W NP_072047.4
XM_011514797.1 1957 Missense Mutation CGG,TGG R511W XP_011513099.1
XM_011514798.2 1957 Missense Mutation CGG,TGG R511W XP_011513100.1
XM_011514799.1 1957 Missense Mutation CGG,TGG R511W XP_011513101.1
XM_017011177.1 1957 Missense Mutation CGG,TGG R578W XP_016866666.1
XM_017011178.1 1957 Missense Mutation CGG,TGG R453W XP_016866667.1
XM_017011179.1 1957 Intron XP_016866668.1

View Full Product Details