Product Details
- SNP ID
-
rs138107415
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:91941140 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGACAGAAGAAGCTGGAGGCCGGCA[A/C]AGCCAAGGTAGGAGAGCCCGAGGCA
- Phenotype
-
MIM: 604001
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
AKAP9
PubMed Links
Gene Details
- Gene
- AKAP9
- Gene Name
- A-kinase anchoring protein 9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005751.4 |
274 |
Missense Mutation |
AAA,ACA |
K14T |
NP_005742.4 |
NM_147185.2 |
274 |
Missense Mutation |
AAA,ACA |
K14T |
NP_671714.1 |
XM_017011642.1 |
274 |
Missense Mutation |
AAA,ACA |
K14T |
XP_016867131.1 |
XM_017011643.1 |
274 |
Missense Mutation |
AAA,ACA |
K14T |
XP_016867132.1 |
XM_017011644.1 |
274 |
Missense Mutation |
AAA,ACA |
K14T |
XP_016867133.1 |
XM_017011645.1 |
274 |
Missense Mutation |
AAA,ACA |
K14T |
XP_016867134.1 |
XM_017011646.1 |
274 |
Missense Mutation |
AAA,ACA |
K14T |
XP_016867135.1 |
XM_017011647.1 |
274 |
Missense Mutation |
AAA,ACA |
K14T |
XP_016867136.1 |
XM_017011648.1 |
274 |
Missense Mutation |
AAA,ACA |
K14T |
XP_016867137.1 |
XM_017011649.1 |
274 |
Missense Mutation |
AAA,ACA |
K14T |
XP_016867138.1 |
XM_017011650.1 |
274 |
Missense Mutation |
AAA,ACA |
K14T |
XP_016867139.1 |
XM_017011651.1 |
274 |
Missense Mutation |
AAA,ACA |
K14T |
XP_016867140.1 |
XM_017011652.1 |
274 |
Missense Mutation |
AAA,ACA |
K14T |
XP_016867141.1 |
XM_017011653.1 |
274 |
Missense Mutation |
AAA,ACA |
K14T |
XP_016867142.1 |
XM_017011654.1 |
274 |
Missense Mutation |
AAA,ACA |
K14T |
XP_016867143.1 |
XM_017011655.1 |
274 |
Intron |
|
|
XP_016867144.1 |
XM_017011656.1 |
274 |
Intron |
|
|
XP_016867145.1 |
XM_017011657.1 |
274 |
Intron |
|
|
XP_016867146.1 |
XM_017011658.1 |
274 |
Intron |
|
|
XP_016867147.1 |
XM_017011659.1 |
274 |
Intron |
|
|
XP_016867148.1 |
XM_017011660.1 |
274 |
Intron |
|
|
XP_016867149.1 |
View Full Product Details