Product Details

SNP ID
rs139104920
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99506061 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGTTGGAATGATAATGACCGAAT[C/T]CCGAGAAGTTATAGACTTAGACCCC
Phenotype
MIM: 611272
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZKSCAN5 PubMed Links

Gene Details

Gene
ZKSCAN5
Gene Name
zinc finger with KRAB and SCAN domains 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318082.1 147 Missense Mutation TCC,TTC S6F NP_001305011.1
NM_001318083.1 147 Missense Mutation TCC,TTC S6F NP_001305012.1
NM_001318084.1 147 UTR 5 NP_001305013.1
NM_014569.3 147 Missense Mutation TCC,TTC S6F NP_055384.1
NM_145102.3 147 Missense Mutation TCC,TTC S6F NP_659570.1
XM_011515999.2 147 Intron XP_011514301.1
XM_017011918.1 147 Missense Mutation TCC,TTC S6F XP_016867407.1
XM_017011919.1 147 Missense Mutation TCC,TTC S6F XP_016867408.1
XM_017011920.1 147 Missense Mutation TCC,TTC S6F XP_016867409.1
XM_017011921.1 147 Missense Mutation TCC,TTC S6F XP_016867410.1
XM_017011922.1 147 Missense Mutation TCC,TTC S6F XP_016867411.1
XM_017011923.1 147 Missense Mutation TCC,TTC S6F XP_016867412.1
XM_017011924.1 147 Missense Mutation TCC,TTC S6F XP_016867413.1
XM_017011925.1 147 Intron XP_016867414.1
XM_017011926.1 147 UTR 5 XP_016867415.1
XM_017011927.1 147 UTR 5 XP_016867416.1
Gene
ZNF394
Gene Name
zinc finger protein 394
There are no transcripts associated with this gene.

View Full Product Details