Product Details

SNP ID
rs139169038
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:88279429 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTGAGTTCCTTTCCCAGCCCTGG[C/T]GGATCCTTGTAAGGGTGTTGTCTAC
Phenotype
MIM: 611098
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STEAP4 PubMed Links

Gene Details

Gene
STEAP4
Gene Name
STEAP4 metalloreductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205315.1 1572 Missense Mutation NP_001192244.1
NM_001205316.1 1572 Missense Mutation NP_001192245.1
NM_024636.3 1572 Missense Mutation NP_078912.2

View Full Product Details