Product Details

SNP ID
rs139313450
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:144095800 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCTTGAGGATCCAGTCTGGGGA[A/G]GGCCGCAGAAAGGCCTTCTCTACCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR2A12 PubMed Links

Gene Details

Gene
OR2A12
Gene Name
olfactory receptor family 2 subfamily A member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004135.1 693 Silent Mutation GAA,GAG E231E NP_001004135.1

View Full Product Details