Product Details

SNP ID
rs139432346
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:88794435 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTGTGTGAGAAAATCTGTGATGA[A/C]CTCCTTTAGGGACTGTGACTGTGTC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C7orf62 PubMed Links

Gene Details

Gene
C7orf62
Gene Name
chromosome 7 open reading frame 62
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152706.3 690 Missense Mutation GTC,TTC V170F NP_689919.1
XM_011515904.2 690 Missense Mutation GTC,TTC V170F XP_011514206.1
Gene
ZNF804B
Gene Name
zinc finger protein 804B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181646.3 690 Intron NP_857597.1

View Full Product Details