Product Details

SNP ID
rs139523163
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100155806 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCAGCCTGCATGGCCCGGCGCTC[C/T]TCCTCACACAGCTGCTTTCCTGGGG
Phenotype
MIM: 608235
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C7orf43 PubMed Links

Gene Details

Gene
C7orf43
Gene Name
chromosome 7 open reading frame 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303470.1 982 Silent Mutation GAA,GAG E151E NP_001290399.1
NM_018275.4 982 Silent Mutation GAA,GAG E420E NP_060745.3
Gene
GAL3ST4
Gene Name
galactose-3-O-sulfotransferase 4
There are no transcripts associated with this gene.

Gene
LAMTOR4
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 4
There are no transcripts associated with this gene.

Gene
MIR4658
Gene Name
microRNA 4658
There are no transcripts associated with this gene.

View Full Product Details