Product Details

SNP ID
rs139805699
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100430865 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGCCGGAGGGAGTGAGAGTCAC[C/T]CCGTGCCGCCCACAGCCCCTCTCAC
Phenotype
MIM: 611478
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
MEPCE PubMed Links
Additional Information
For this assay, SNP(s) [rs6962151] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
MEPCE
Gene Name
methylphosphate capping enzyme
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001194990.1 502 UTR 5 NP_001181919.1
NM_001194991.1 502 Intron NP_001181920.1
NM_001194992.1 502 Intron NP_001181921.1
NM_019606.5 502 Missense Mutation CCC,TCC P283S NP_062552.2
XM_011516410.2 502 Intron XP_011514712.1
Gene
PPP1R35
Gene Name
protein phosphatase 1 regulatory subunit 35
There are no transcripts associated with this gene.

Gene
ZCWPW1
Gene Name
zinc finger CW-type and PWWP domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258008.1 502 Intron NP_001244937.1
NM_017984.4 502 Intron NP_060454.3

View Full Product Details