Product Details

SNP ID
rs140252124
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:88794777 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTTCTTTCTACATTTGCTTGTA[C/T]CTTGGCCACTAGAAACATCCTATCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C7orf62 PubMed Links

Gene Details

Gene
C7orf62
Gene Name
chromosome 7 open reading frame 62
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152706.3 348 Missense Mutation ATA,GTA I56V NP_689919.1
XM_011515904.2 348 Missense Mutation ATA,GTA I56V XP_011514206.1
Gene
ZNF804B
Gene Name
zinc finger protein 804B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181646.3 348 Intron NP_857597.1

View Full Product Details