Product Details

SNP ID
rs140475462
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:149474633 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACGGTGCAGGTGAAGGGCCGCAC[A/G]CCCGTGTGCGTTCGGTAGTGGTCGA
Phenotype
MIM: 613914
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ZNF746 PubMed Links

Gene Details

Gene
ZNF746
Gene Name
zinc finger protein 746
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163474.1 2087 Silent Mutation GGC,GGT G563G NP_001156946.1
NM_152557.4 2087 Silent Mutation GGC,GGT G562G NP_689770.3
XM_005249955.2 2087 Silent Mutation GGC,GGT G578G XP_005250012.1
XM_005249956.2 2087 Silent Mutation GGC,GGT G577G XP_005250013.1
XM_005249957.1 2087 Silent Mutation GGC,GGT G565G XP_005250014.1
XM_011515838.1 2087 Silent Mutation GGC,GGT G550G XP_011514140.1

View Full Product Details