Product Details

SNP ID
rs140695705
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:77794097 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTGTTCCAGTTTAGCACAGAGTG[C/T]GAGGCAGAAGGCTAATTTAAGTAAC
Phenotype
MIM: 616785
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PHTF2 PubMed Links

Gene Details

Gene
PHTF2
Gene Name
putative homeodomain transcription factor 2
There are no transcripts associated with this gene.

Gene
TMEM60
Gene Name
transmembrane protein 60
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032936.3 504 Missense Mutation ACA,GCA T93A NP_116325.1

View Full Product Details