Product Details

SNP ID
rs140828281
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:143391625 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACAGCTCCAGCTCCTTACTCAGC[A/G]GTCAGCTCCAGCACACACTCCATGG
Phenotype
MIM: 179610 MIM: 602002
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EPHA1 PubMed Links

Gene Details

Gene
EPHA1
Gene Name
EPH receptor A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005232.4 2934 Silent Mutation ACC,ACT T949T NP_005223.4
XM_006715880.3 2934 Silent Mutation ACC,ACT T834T XP_006715943.1
Gene
FAM131B
Gene Name
family with sequence similarity 131 member B
There are no transcripts associated with this gene.

Gene
MIR6892
Gene Name
microRNA 6892
There are no transcripts associated with this gene.

Gene
ZYX
Gene Name
zyxin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010972.1 2934 Intron NP_001010972.1
NM_003461.4 2934 Intron NP_003452.1
XM_011516569.2 2934 Intron XP_011514871.2
XM_017012587.1 2934 Intron XP_016868076.1

View Full Product Details