Product Details

SNP ID
rs140932994
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:44582104 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAAACAGCGCTTCTCGGTCTCGC[C/G]GATGTGGAAGTAGAGCCCCTGGGCG
Phenotype
MIM: 608023 MIM: 612038
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DDX56 PubMed Links

Gene Details

Gene
DDX56
Gene Name
DEAD-box helicase 56
There are no transcripts associated with this gene.

Gene
TMED4
Gene Name
transmembrane p24 trafficking protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303058.1 192 Missense Mutation CGC,GGC R35G NP_001289987.1
NM_001303059.1 192 Missense Mutation CGC,GGC R35G NP_001289988.1
NM_001303060.1 192 UTR 5 NP_001289989.1
NM_001303061.1 192 UTR 5 NP_001289990.1
NM_001303062.1 192 UTR 5 NP_001289991.1
NM_182547.3 192 Missense Mutation CGC,GGC R35G NP_872353.2

View Full Product Details