Product Details

SNP ID
rs141056346
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:143026458 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCTTTCCTCCGGCCAGAGGCTGA[C/T]GGGATCTTGAGGATGGTGGAGATAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR9A2 PubMed Links

Gene Details

Gene
OR9A2
Gene Name
olfactory receptor family 9 subfamily A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001658.1 675 Silent Mutation CCA,CCG P225P NP_001001658.1

View Full Product Details