Product Details

SNP ID
rs141718556
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:113877979 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATTCCTTTTCACCTGAAGTGTAT[A/G]GTGATTGCCCAGAGCTTACTAATCC
Phenotype
MIM: 600917
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPP1R3A PubMed Links

Gene Details

Gene
PPP1R3A
Gene Name
protein phosphatase 1 regulatory subunit 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002711.3 2690 Missense Mutation CCA,CTA P1038L NP_002702.2
XM_005250473.3 2690 Missense Mutation CCA,CTA P837L XP_005250530.1

View Full Product Details