Product Details

SNP ID
rs141942249
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:107203605 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTGCCTTCTCTACTTCTCACTG[A/G]TTGAACATAAGCTTCCAGGGCTCCC
Phenotype
MIM: 606821 MIM: 616714
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COG5 PubMed Links

Gene Details

Gene
COG5
Gene Name
component of oligomeric golgi complex 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161520.1 3019 Intron NP_001154992.1
NM_006348.3 3019 Missense Mutation NP_006339.3
NM_181733.2 3019 Missense Mutation NP_859422.2
Gene
HBP1
Gene Name
HMG-box transcription factor 1
There are no transcripts associated with this gene.

View Full Product Details