Product Details

SNP ID
rs142171905
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:152136741 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAAAAGTCATAAAACAGAAAACA[A/G]AAATCTCTTTCTGTCTGCAAAATTC
Phenotype
MIM: 606833
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KMT2C PubMed Links

Gene Details

Gene
KMT2C
Gene Name
lysine methyltransferase 2C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_170606.2 15241 UTR 3 NP_733751.2
XM_005250025.4 15241 UTR 3 XP_005250082.1
XM_005250026.3 15241 UTR 3 XP_005250083.1
XM_005250027.4 15241 UTR 3 XP_005250084.1
XM_005250028.4 15241 UTR 3 XP_005250085.1
XM_005250031.4 15241 UTR 3 XP_005250088.1
XM_006716077.3 15241 UTR 3 XP_006716140.1
XM_006716078.3 15241 UTR 3 XP_006716141.1
XM_006716079.3 15241 UTR 3 XP_006716142.1
XM_011516450.2 15241 UTR 3 XP_011514752.1
XM_011516451.2 15241 UTR 3 XP_011514753.1
XM_011516452.2 15241 UTR 3 XP_011514754.1
XM_011516453.2 15241 UTR 3 XP_011514755.1
XM_011516454.2 15241 UTR 3 XP_011514756.1
XM_011516456.2 15241 UTR 3 XP_011514758.1
XM_017012480.1 15241 UTR 3 XP_016867969.1
XM_017012481.1 15241 UTR 3 XP_016867970.1
XM_017012482.1 15241 UTR 3 XP_016867971.1
XM_017012483.1 15241 UTR 3 XP_016867972.1
XM_017012484.1 15241 UTR 3 XP_016867973.1
XM_017012485.1 15241 UTR 3 XP_016867974.1
XM_017012486.1 15241 UTR 3 XP_016867975.1
XM_017012487.1 15241 UTR 3 XP_016867976.1
XM_017012488.1 15241 UTR 3 XP_016867977.1
XM_017012489.1 15241 UTR 3 XP_016867978.1
XM_017012490.1 15241 UTR 3 XP_016867979.1
Gene
LOC731075
Gene Name
uncharacterized LOC731075
There are no transcripts associated with this gene.

Gene
YBX1P4
Gene Name
Y-box binding protein 1 pseudogene 4
There are no transcripts associated with this gene.

View Full Product Details