Product Details

SNP ID
rs142461818
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:101240905 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCTTGCTCCCTTTGGGCAGCAG[C/T]TCTGGGGAGGGGCAGAGAAGAGGGT
Phenotype
MIM: 615778 MIM: 609003
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLDN15 PubMed Links

Gene Details

Gene
CLDN15
Gene Name
claudin 15
There are no transcripts associated with this gene.

Gene
FIS1
Gene Name
fission, mitochondrial 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016068.2 266 Silent Mutation GAA,GAG E60E NP_057152.2

View Full Product Details