Product Details

SNP ID
rs142606351
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100156636 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCCTCCAGGATCCACACTCACCG[A/G]TAGTGGGTGTAGATGCTCTGAGTGA
Phenotype
MIM: 608235
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C7orf43 PubMed Links

Gene Details

Gene
C7orf43
Gene Name
chromosome 7 open reading frame 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303470.1 796 Silent Mutation TAC,TAT Y89Y NP_001290399.1
NM_018275.4 796 Silent Mutation TAC,TAT Y358Y NP_060745.3
Gene
GAL3ST4
Gene Name
galactose-3-O-sulfotransferase 4
There are no transcripts associated with this gene.

Gene
LAMTOR4
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 4
There are no transcripts associated with this gene.

Gene
MIR4658
Gene Name
microRNA 4658
There are no transcripts associated with this gene.

View Full Product Details