Product Details

SNP ID
rs143394460
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:144095379 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCTTGTGATGCACAAAAAAGTCA[A/T]CTCCTTTGCTCCTTGCATACTTCAG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR2A12 PubMed Links

Gene Details

Gene
OR2A12
Gene Name
olfactory receptor family 2 subfamily A member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004135.1 272 Missense Mutation AAC,ATC N91I NP_001004135.1

View Full Product Details