Product Details

SNP ID
rs143452198
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:1542694 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGATTTTTTTATTTAAGAAAAAGA[C/T]GAGGGACTCTTTGTCACGTGGGTTT
Phenotype
MIM: 600197
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100128653 PubMed Links

Gene Details

Gene
LOC100128653
Gene Name
uncharacterized LOC100128653
There are no transcripts associated with this gene.

Gene
MAFK
Gene Name
MAF bZIP transcription factor K
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002360.3 5817 UTR 3 NP_002351.1
XM_005249851.2 5817 Intron XP_005249908.2
XM_006715773.2 5817 UTR 3 XP_006715836.1
Gene
TMEM184A
Gene Name
transmembrane protein 184A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001097620.1 5817 UTR 3 NP_001091089.1
XM_006715653.1 5817 Intron XP_006715716.1
XM_006715655.2 5817 Intron XP_006715718.1
XM_011515164.1 5817 Intron XP_011513466.1
XM_011515165.2 5817 Intron XP_011513467.1
XM_011515166.2 5817 Intron XP_011513468.1
XM_017011815.1 5817 Intron XP_016867304.1

View Full Product Details