Product Details

SNP ID
rs143534989
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:95405372 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCTCTGTGGTATAAAGTGCCTAT[A/G]AGCAGCTTCCCATCATACACTGAGG
Phenotype
MIM: 602447 MIM: 602720
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PON2 PubMed Links

Gene Details

Gene
PON2
Gene Name
paraoxonase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000305.2 1019 Silent Mutation CTC,CTT L341L NP_000296.2
NM_001018161.1 1019 Silent Mutation CTC,CTT L329L NP_001018171.1
XM_005250453.1 1019 Silent Mutation CTC,CTT L273L XP_005250510.1
XM_017012357.1 1019 Silent Mutation CTC,CTT L271L XP_016867846.1
XM_017012358.1 1019 Silent Mutation CTC,CTT L255L XP_016867847.1
Gene
PON3
Gene Name
paraoxonase 3
There are no transcripts associated with this gene.

View Full Product Details