Product Details

SNP ID
rs143868420
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:120788728 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGAGTTGTCATTCTTCAAGGAC[A/G]TCATTTGGTCTGTCCCCGGCTCCCT
Phenotype
MIM: 613138
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TSPAN12 PubMed Links

Gene Details

Gene
TSPAN12
Gene Name
tetraspanin 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012338.3 663 Missense Mutation ACG,ATG T261M NP_036470.1
XM_005250239.2 663 Missense Mutation ACG,ATG T261M XP_005250296.1
XM_011515993.1 663 Missense Mutation ACG,ATG T261M XP_011514295.1
XM_011515994.1 663 Missense Mutation ACG,ATG T261M XP_011514296.1
XM_017011912.1 663 Missense Mutation ACG,ATG T261M XP_016867401.1
XM_017011913.1 663 Missense Mutation ACG,ATG T236M XP_016867402.1
XM_017011914.1 663 Missense Mutation ACG,ATG T134M XP_016867403.1

View Full Product Details