Product Details

SNP ID
rs144294259
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:55953299 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGTGAGAGTGACCAGGCTTGTTC[G/T]GGATCCCTATTTATTAAAGGTGAGA
Phenotype
MIM: 611980
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MRPS17 PubMed Links

Gene Details

Gene
MRPS17
Gene Name
mitochondrial ribosomal protein S17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015969.2 Intron NP_057053.1

View Full Product Details