Product Details
- SNP ID
-
rs144353491
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:1233621 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAGGAGCTGGAGAAGGCGTTCAAC[A/G]AGAGCCACTATCCCGACGTGTTCAT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
UNCX
PubMed Links
Gene Details
- Gene
- UNCX
- Gene Name
- UNC homeobox
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001080461.1 |
376 |
Missense Mutation |
AAG,GAG |
K126E |
NP_001073930.1 |
View Full Product Details