Product Details

SNP ID
rs144761422
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:76003537 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGAGCCTTGGGCCCCACTGCCC[C/T]CGAGGTAAACACCACGGTTGCTCGT
Phenotype
MIM: 616695 MIM: 616550
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STYXL1 PubMed Links

Gene Details

Gene
STYXL1
Gene Name
serine/threonine/tyrosine interacting-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317785.1 Intron NP_001304714.1
NM_001317786.1 Intron NP_001304715.1
NM_001317787.1 Intron NP_001304716.1
NM_001317788.1 Intron NP_001304717.1
NM_001317789.1 Intron NP_001304718.1
NM_016086.2 Intron NP_057170.1
XM_011516292.2 Intron XP_011514594.1
XM_011516293.2 Intron XP_011514595.1
XM_011516300.1 Intron XP_011514602.1
XM_017012296.1 Intron XP_016867785.1
XM_017012297.1 Intron XP_016867786.1
XM_017012298.1 Intron XP_016867787.1
XM_017012299.1 Intron XP_016867788.1
XM_017012300.1 Intron XP_016867789.1
XM_017012301.1 Intron XP_016867790.1
XM_017012302.1 Intron XP_016867791.1
Gene
TMEM120A
Gene Name
transmembrane protein 120A
There are no transcripts associated with this gene.

View Full Product Details