Product Details

SNP ID
rs144844975
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:73691062 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGTTAGCTGCCTGTCCCTCCCT[A/G]GCACTGCAGGTAGAGTGAGGAAATT
Phenotype
MIM: 186590 MIM: 615733
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJC30 PubMed Links

Gene Details

Gene
DNAJC30
Gene Name
DnaJ heat shock protein family (Hsp40) member C30
There are no transcripts associated with this gene.

Gene
STX1A
Gene Name
syntaxin 1A
There are no transcripts associated with this gene.

Gene
WBSCR22
Gene Name
Williams-Beuren syndrome chromosome region 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202560.2 Intron NP_001189489.1
NM_017528.4 Intron NP_059998.2
XM_006715847.1 Intron XP_006715910.1
XM_011515778.1 Intron XP_011514080.1
XM_011515779.1 Intron XP_011514081.1

View Full Product Details