Product Details

SNP ID
rs144914171
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:27147501 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGAGGGCGAGGCGCCACTGAGGTC[C/T]TTATCAGAATAGAAACACGAGGCCC
Phenotype
MIM: 142954 MIM: 142952 MIM: 142951 MIM: 142950
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HOXA-AS3 PubMed Links

Gene Details

Gene
HOXA-AS3
Gene Name
HOXA cluster antisense RNA 3
There are no transcripts associated with this gene.

Gene
HOXA3
Gene Name
homeobox A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030661.4 274 Intron NP_109377.1
NM_153631.2 274 Intron NP_705895.1
XM_005249730.2 274 Intron XP_005249787.1
XM_005249731.2 274 Intron XP_005249788.1
XM_005249732.3 274 Intron XP_005249789.1
XM_006715715.2 274 Intron XP_006715778.1
XM_011515343.2 274 Intron XP_011513645.1
Gene
HOXA5
Gene Name
homeobox A5
There are no transcripts associated with this gene.

Gene
HOXA6
Gene Name
homeobox A6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024014.3 274 Silent Mutation AAA,AAG K83K NP_076919.1
Gene
HOXA7
Gene Name
homeobox A7
There are no transcripts associated with this gene.

View Full Product Details