Product Details

SNP ID
rs145174585
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:19145349 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCGTAGTGTCCACGCAGCTCTCC[C/G]GATAGGCCGCCATCGCTTCGGCTTG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FERD3L PubMed Links

Gene Details

Gene
FERD3L
Gene Name
Fer3-like bHLH transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152898.2 73 Missense Mutation CCG,CGG P5R NP_690862.1

View Full Product Details