Product Details

SNP ID
rs145550034
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:150801621 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACACCCACATCGATGTGCACATCC[A/G]CCAGGAGTCTGCCCTGGCCAAGCTC
Phenotype
MIM: 610334 MIM: 610385
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM176A PubMed Links

Gene Details

Gene
TMEM176A
Gene Name
transmembrane protein 176A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018487.2 284 Missense Mutation CAC,CGC H24R NP_060957.2
XM_011516376.2 284 Missense Mutation CAC,CGC H41R XP_011514678.1
XM_011516377.2 284 Missense Mutation CAC,CGC H41R XP_011514679.1
XM_011516378.2 284 Missense Mutation CAC,CGC H41R XP_011514680.1
XM_017012393.1 284 Missense Mutation CAC,CGC H24R XP_016867882.1
Gene
TMEM176B
Gene Name
transmembrane protein 176B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101311.1 284 Intron NP_001094781.1
NM_001101312.1 284 Intron NP_001094782.1
NM_001101314.1 284 Intron NP_001094784.1
NM_014020.3 284 Intron NP_054739.3
XM_006715933.3 284 Intron XP_006715996.2

View Full Product Details