Product Details
- SNP ID
-
rs145632958
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:6023498 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTAAAGAAAAAGCCGCTGTTTTCC[A/G]CTCCATGAACTCTGCTCTTGGGAAG
- Phenotype
-
MIM: 600859
MIM: 613635
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AIMP2
PubMed Links
Gene Details
- Gene
- AIMP2
- Gene Name
- aminoacyl tRNA synthetase complex interacting multifunctional protein 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001326606.1 |
763 |
Missense Mutation |
CAC,CGC |
H217R |
NP_001313535.1 |
NM_001326607.1 |
763 |
Missense Mutation |
CAC,CGC |
H188R |
NP_001313536.1 |
NM_001326609.1 |
763 |
Missense Mutation |
CAC,CGC |
H179R |
NP_001313538.1 |
NM_001326611.1 |
763 |
Missense Mutation |
CAC,CGC |
H179R |
NP_001313540.1 |
NM_006303.3 |
763 |
Missense Mutation |
CAC,CGC |
H257R |
NP_006294.2 |
XM_005249847.3 |
763 |
Missense Mutation |
CAC,CGC |
H250R |
XP_005249904.1 |
XM_005249848.2 |
763 |
Missense Mutation |
CAC,CGC |
H217R |
XP_005249905.1 |
XM_005249850.3 |
763 |
Missense Mutation |
CAC,CGC |
H179R |
XP_005249907.1 |
XM_006715772.2 |
763 |
Missense Mutation |
CAC,CGC |
H213R |
XP_006715835.1 |
XM_011515519.1 |
763 |
Missense Mutation |
CAC,CGC |
H228R |
XP_011513821.1 |
- Gene
- ANKRD61
- Gene Name
- ankyrin repeat domain 61
There are no transcripts associated with this gene.
- Gene
- EIF2AK1
- Gene Name
- eukaryotic translation initiation factor 2 alpha kinase 1
View Full Product Details