Product Details

SNP ID
rs146389256
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:39566442 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATGTCGTGGGTTCAAGCAGCCTC[C/T]TTGATCCAGGGCCCTGGAGACAAAG
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
LOC105375240 PubMed Links

Gene Details

Gene
LOC105375240
Gene Name
uncharacterized LOC105375240
There are no transcripts associated with this gene.

Gene
YAE1D1
Gene Name
Yae1 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282446.1 67 Silent Mutation TCC,TCT S8S NP_001269375.1
NM_020192.4 67 Silent Mutation TCC,TCT S8S NP_064577.1

View Full Product Details