Product Details

SNP ID
rs146984976
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:87940700 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAAGTAATTATATTTGACTGTGG[A/G]TAGCCTTGCTATCTTCAAAGTTGTT
Phenotype
MIM: 603709
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAM22 PubMed Links

Gene Details

Gene
ADAM22
Gene Name
ADAM metallopeptidase domain 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324417.1 Intron NP_001311346.1
NM_001324418.1 Intron NP_001311347.1
NM_001324419.1 Intron NP_001311348.1
NM_001324420.1 Intron NP_001311349.1
NM_001324421.1 Intron NP_001311350.1
NM_004194.4 Intron NP_004185.1
NM_016351.5 Intron NP_057435.2
NM_021721.4 Intron NP_068367.1
NM_021722.5 Intron NP_068368.2
NM_021723.4 Intron NP_068369.1
XM_005250445.3 Intron XP_005250502.1
XM_006716028.3 Intron XP_006716091.1
XM_006716029.3 Intron XP_006716092.1
XM_011516318.1 Intron XP_011514620.1
XM_011516319.2 Intron XP_011514621.1
XM_011516320.2 Intron XP_011514622.1
XM_011516321.2 Intron XP_011514623.1
XM_011516322.1 Intron XP_011514624.1
XM_011516323.1 Intron XP_011514625.1
XM_011516324.1 Intron XP_011514626.1
XM_017012329.1 Intron XP_016867818.1
XM_017012330.1 Intron XP_016867819.1
XM_017012331.1 Intron XP_016867820.1
XM_017012332.1 Intron XP_016867821.1
XM_017012333.1 Intron XP_016867822.1
XM_017012334.1 Intron XP_016867823.1
XM_017012335.1 Intron XP_016867824.1
XM_017012336.1 Intron XP_016867825.1
XM_017012337.1 Intron XP_016867826.1
XM_017012338.1 Intron XP_016867827.1
XM_017012339.1 Intron XP_016867828.1
XM_017012340.1 Intron XP_016867829.1
XM_017012341.1 Intron XP_016867830.1

View Full Product Details